accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)(S)(S)(S)(R)(S)(R)(R)OO(R)OH(R)HOHHHHNaMeMeMe

CAS RN 302-95-4

IUPAC Name: sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
Synonyms: sodium deoxycholate|sodium desoxycholate|deoxycholate sodium|deoxycholic acid sodium salt|desoxycholate sodium|sodium 7-deoxycholate
Molecular Weight (g/mol): 414.56
Molecular Formula: C24H39NaO4
InChi Key: FHHPUSMSKHSNKW-SMOYURAASA-M
SMILES: [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
Form 
  • (3)
Percent Purity 
  • (1)
  • (2)
Product Type 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results