Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxycholic Acid Sodium Salt, Fisher BioReagents

Catalog No. BP349100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP349-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP349-100

missing translation for 'validMainframeMsg'

For solubilizing and isolating membrane-bound proteins in an active state

This anionic detergent is useful for solubilizing and isolating membrane-bound proteins in an active state.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-95-4
Molecular Formula C24H39NaO4
Molecular Weight (g/mol) 414.56
MDL Number MFCD00064139
InChI Key FHHPUSMSKHSNKW-SMOYURAASA-M
Synonym Sodium Deoxycholate, 3-alpha, 12-alpha-Dihydroxy-5-beta-cholan-24-oic acid, sodium salt
PubChem CID 91810855
IUPAC Name sodium;(4R)-4-[(3R,5R,8R,10S,12S,13R,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Color White
Physical Form Powder/Solid
Melting Point 357°C
pH 7.5 to 10
Quantity 100 g
Identification Pass Test
Packaging Poly Bottle
Assay Percent Range ≥99 %
Formula Weight 414.58
Percent Purity ≥99%
Chemical Name or Material Deoxycholic Acid Sodium Salt
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.