accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30216-44-5

IUPAC Name: 2-bromobenzene-1-carbothioamide
Synonyms: 2-bromobenzenecarbothioamide
Molecular Weight (g/mol): 216.1
Molecular Formula: C7H6BrNS
InChi Key: GHCQWSAFBXLYSO-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results