Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromothiobenzamide, 98%, Thermo Scientific™

Catalog No. AC374320010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC374320010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC374320010 Supplier Thermo Scientific Chemicals Supplier No. 374320010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30216-44-5
Molecular Formula C7H6BrNS
Molecular Weight (g/mol) 216.096
InChI Key GHCQWSAFBXLYSO-UHFFFAOYSA-N
Synonym 2-bromothiobenzamide, 2-bromobenzothioamide, thiobenzamide, 2-bromo, 2-bromobenzene-1-carbothioamide, benzenecarbothioamide,2-bromo, amino 2-bromophenyl methane-1-thione, 2-bromo-thiobenzamide, acmc-1agnp
PubChem CID 2734818
IUPAC Name 2-bromobenzenecarbothioamide
SMILES C1=CC=C(C(=C1)C(=S)N)Br

Specifications

Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 216.1
Chemical Name or Material 2-Bromothiobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.