accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3022-16-0

IUPAC Name: 1,4-bis(chloromethyl)-2,3,5,6-tetramethylbenzene
Synonyms: 1,4-bis(chloromethyl)-2,3,5,6-tetramethylbenzene
Molecular Weight (g/mol): 231.16
Molecular Formula: C12H16Cl2
InChi Key: PGFAKOSRZYDFLR-UHFFFAOYSA-N
SMILES: CC1=C(C)C(CCl)=C(C)C(C)=C1CCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results