Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Bis(chloromethyl)durene 98.0+%, TCI America™
SDP

Catalog No. B17835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17835G Supplier TCI America Supplier No. B17835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3022-16-0
Molecular Formula C12H16Cl2
Molecular Weight (g/mol) 231.16
MDL Number MFCD00018883
InChI Key PGFAKOSRZYDFLR-UHFFFAOYSA-N
Synonym 1,4-Bis(chloromethyl)-2,3,5,6-tetramethylbenzene
PubChem CID 76402
IUPAC Name 1,4-bis(chloromethyl)-2,3,5,6-tetramethylbenzene
SMILES CC1=C(C(=C(C(=C1CCl)C)C)CCl)C

Specifications

Melting Point 196°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 231.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Bis(chloromethyl)durene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.