CAS RN 30269-04-6
IUPAC Name:
pyrene-1,8-diamine
Synonyms:
pyrene-1,8-diamine
Molecular Weight (g/mol):
232.29
Molecular Formula:
C16H12N2
InChi Key:
BLYOXQBERINFDU-UHFFFAOYSA-N
SMILES:
NC1=CC=C2C=CC3=CC=C(N)C4=CC=C1C2=C34
1
–
1
of
1
results
1
1,8-Diaminopyrene 97.0+%, TCI America™
CAS: 30269-04-6 Molecular Formula: C16H12N2 Molecular Weight (g/mol): 232.286 MDL Number: MFCD00142575 InChI Key: BLYOXQBERINFDU-UHFFFAOYSA-N PubChem CID: 169139 IUPAC Name: pyrene-1,8-diamine SMILES: C1=CC2=C3C(=C(C=C2)N)C=CC4=C(C=CC1=C43)N