Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diaminopyrene 97.0+%, TCI America™
SDP

Catalog No. D2168100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2168100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2168100MG Supplier TCI America Supplier No. D2168100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30269-04-6
Molecular Formula C16H12N2
Molecular Weight (g/mol) 232.286
MDL Number MFCD00142575
InChI Key BLYOXQBERINFDU-UHFFFAOYSA-N
PubChem CID 169139
IUPAC Name pyrene-1,8-diamine
SMILES C1=CC2=C3C(=C(C=C2)N)C=CC4=C(C=CC1=C43)N

Specifications

Melting Point 180°C
Color Green-Yellow
Quantity 100 mg
Formula Weight 232.29
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Diaminopyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.