accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NH2

CAS RN 30273-11-1

IUPAC Name: 4-(butan-2-yl)aniline
Synonyms: 4-(sec-butyl)aniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: NVVVQTNTLIAISI-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results