Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-sec-Butylaniline 98.0+%, TCI America™
SDP

Catalog No. B12565ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12565ML 5 mL
B125625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12565ML Supplier TCI America Supplier No. B12565ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30273-11-1
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00010223
InChI Key NVVVQTNTLIAISI-UHFFFAOYNA-N
PubChem CID 121771
IUPAC Name 4-(butan-2-yl)aniline
SMILES CCC(C)C1=CC=C(N)C=C1

Specifications

Color Yellow
Boiling Point 238°C
Quantity 5 mL
UN Number 2810
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-sec-Butylaniline
EINECSNumber (3)-0206
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.