accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 302841-86-7

IUPAC Name: ethyl N-(2,2-diphenylacetyl)carbamate
Synonyms: ethyl N-(2,2-diphenylacetyl)carbamate
Molecular Weight (g/mol): 283.33
Molecular Formula: C17H17NO3
InChi Key: ILSZPWZFQHSKLW-UHFFFAOYSA-N
SMILES: CCOC(=O)NC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results