accessibility menu, dialog, popup

UserName

Viewing profile as user:
NCOFFFF

CAS RN 302912-19-2

IUPAC Name: 1-fluoro-3-isocyanato-5-(trifluoromethyl)benzene
Synonyms: 1-fluoro-3-isocyanato-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 205.11
Molecular Formula: C8H3F4NO
InChi Key: WGSXKYXKAARAKD-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(=C1)N=C=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results