Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-(trifluoromethyl)phenyl isocyanate, Thermo Scientific™

Catalog No. MAY00117DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00117DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00117DA Supplier Thermo Scientific Chemicals Supplier No. MAY00117DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 302912-19-2
Molecular Formula C8H3F4NO
Molecular Weight (g/mol) 205.112
InChI Key WGSXKYXKAARAKD-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl phenyl isocyanate, 1-fluoro-3-isocyanato-5-trifluoromethyl benzene, 3-fluoro-5-trifluoromethyl phenyl, 3-fluoro-5-trifluoromethylphenylisocyanate, 3-fluoro-5-trifluoromethylphenyl isocyanate, 3-fluoro-5-trifluoromethyl phenylisocyanate, 5-fluoro-3-trifluoromethyl benzenisocyanate, 3-fluoro-5-trifluoromethyl-phenylisocyanate, benzene,1-fluoro-3-isocyanato-5-trifluoromethyl
PubChem CID 3445672
IUPAC Name 1-fluoro-3-isocyanato-5-(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1N=C=O)F)C(F)(F)F

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.11
Percent Purity 97%
Chemical Name or Material 3-fluoro-5-(trifluoromethyl)phenyl isocyanate
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.