accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHFF

CAS RN 302912-34-1

IUPAC Name: 1-ethynyl-2,4-difluorobenzene
Synonyms: 1-ethynyl-2,4-difluorobenzene
Molecular Weight (g/mol): 138.12
Molecular Formula: C8H4F2
InChi Key: HRUJQXRGWQWYDH-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C=C1)C#C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results