Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethynyl-2,4-difluorobenzene 96.0+%, TCI America™
SDP

Catalog No. E11695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11695G 5 g
E11691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11695G Supplier TCI America Supplier No. E11695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 302912-34-1
Molecular Formula C8H4F2
Molecular Weight (g/mol) 138.117
MDL Number MFCD02093726
InChI Key HRUJQXRGWQWYDH-UHFFFAOYSA-N
Synonym 2,4-Difluorophenylacetylene
PubChem CID 5063820
IUPAC Name 1-ethynyl-2,4-difluorobenzene
SMILES C#CC1=C(C=C(C=C1)F)F

Specifications

Boiling Point 50°C
Quantity 5 g
UN Number 1325
Formula Weight 138.12
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethynyl-2,4-difluorobenzene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.