accessibility menu, dialog, popup

UserName

Viewing profile as user:
SBrBr

CAS RN 302912-44-3

IUPAC Name: 2,5-dibromo-3-cyclohexylthiophene
Synonyms: 2,5-dibromo-3-cyclohexylthiophene
Molecular Weight (g/mol): 324.07
Molecular Formula: C10H12Br2S
InChi Key: GQDHHQWPBULMEB-UHFFFAOYSA-N
SMILES: BrC1=CC(C2CCCCC2)=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results