accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOHHOOOH

CAS RN 303127-80-2

IUPAC Name: 4-(benzyloxy)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,5-diol
Synonyms: 4-(benzyloxy)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,5-diol
Molecular Weight (g/mol): 376.41
Molecular Formula: C20H24O7
InChi Key: CZPFTCBIXZWFIZ-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(CO)C(O)C(OCC3=CC=CC=C3)C2O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results