accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30363-03-2

IUPAC Name: tris(4-bromophenyl)-1,3,5-triazine
Synonyms: tris(4-bromophenyl)-1,3,5-triazine
Molecular Weight (g/mol): 546.06
Molecular Formula: C21H12Br3N3
InChi Key: WZYVDGDZBNQVCF-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results