Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tris(4-bromophenyl)-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. T3178200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3178200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3178200MG Supplier TCI America Supplier No. T3178200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30363-03-2
Molecular Formula C21H12Br3N3
Molecular Weight (g/mol) 546.06
MDL Number MFCD04116312
InChI Key WZYVDGDZBNQVCF-UHFFFAOYSA-N
PubChem CID 35224
IUPAC Name tris(4-bromophenyl)-1,3,5-triazine
SMILES BrC1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 361°C
Color White-Yellow
Quantity 200 mg
Formula Weight 546.06
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris(4-bromophenyl)-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.