accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 304-21-2

IUPAC Name: 7-methoxy-1-methyl-2H,3H,4H-pyrido[3,4-b]indole
Synonyms: harmaline
Molecular Weight (g/mol): 214.27
Molecular Formula: C13H14N2O
InChi Key: QJOZJXNKVMFAET-UHFFFAOYSA-N
SMILES: COC1=CC2=NC3=C(C)NCCC3=C2C=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)