Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harmaline, 98+%

Catalog No. AAJ6169903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6169903 1 g
AAJ6169906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6169903 Supplier Thermo Scientific Chemicals Supplier No. J6169903
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 304-21-2 | C13H14N2O | 214.268 g/mol

CNS stimulant; may act through NMDA receptors. Reversible inhibitor of MAO-A

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • CNS stimulant; may act through NMDA receptors
  • Reversible inhibitor of MAO-A
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304-21-2
Molecular Formula C13H14N2O
Molecular Weight (g/mol) 214.268
MDL Number MFCD00004955
InChI Key QJOZJXNKVMFAET-UHFFFAOYSA-N
Synonym harmaline, dihydroharmine, harmidine, armalin, harmalol methyl ether, o-methylharmalol, 3,4-dihydroharmine, harmine, dihydro, harmalin, 1-methyl-7-methoxy-3,4-dihydro-beta-carboline
PubChem CID 5280951
IUPAC Name 7-methoxy-1-methyl-3,4-dihydro-2H-pyrido[3,4-b]indole
SMILES CC1=C2C(=C3C=CC(=CC3=N2)OC)CCN1

Specifications

Melting Point 239°C to 241°C
Color Brown
Quantity 1 g
Beilstein 207310
Merck Index 14,4613
Formula Weight 214.27
Percent Purity ≥98%
Physical Form Crystals
Chemical Name or Material Harmaline
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 206-152-6
RTECSNumber UU9800000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.