accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30433-91-1

IUPAC Name: 2-(thiophen-2-yl)ethan-1-aminium
Synonyms: 2-(thiophen-2-yl)ethanaminium
Molecular Weight (g/mol): 128.21
Molecular Formula: C6H10NS
InChi Key: HVLUYXIJZLDNIS-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=CS1
Boiling Point 
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)