Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Thiopheneethylamine

Catalog No. 4232705G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-327-05G 5 g
42-327-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-327-05G Supplier Sigma Aldrich Supplier No. 4232705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-thiopheneethylamine (2-(thiophen-2-yl)ethanamine) is an aromatic amine
  • It undergoes microwave induced condensation with iminodiacetic acid to form the corresponding piperazine-2,6-dione derivatives
  • Its effect as a probable substitute to the pyridine ligand on the performance of poly(3-hexylthiophene)/cdse hybrid solar cells has been investigated.2-thiopheneethylamine (2-thiophene ethyl amine, 2-(thien-2-yl)ethylamine) is suitable to functionalize multiwall carbon nanotubes (mwcnt).it may be used as a reactant in the synthesis of pyrimidine derivatives by reacting with various isothiocyanatoketones and acylguanidines derivatives by reacting with aroyl s-methylisothiourea

Specifications

CAS 30433-91-1
Density 1.087 g/mL (at 25°C (literature))
Boiling Point 200°C to 201°C (750 mmHg)
Molecular Formula C6H9NS
Refractive Index n20/D 1.551 (literature)
Linear Formula C6H9NS
MDL Number MFCD00051495
Quantity 5 g
Molecular Weight (g/mol) 127.21
Percent Purity 96%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.