accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHNNH2OOH

CAS RN 304655-78-5

IUPAC Name: 3-amino-1H-1,2,4-triazole-5-carboxylic acid
Synonyms: 5-amino-2H-1,2,4-triazole-3-carboxylic acid
Molecular Weight (g/mol): 128.09
Molecular Formula: C3H4N4O2
InChi Key: MVRGLMCHDCMPKD-UHFFFAOYSA-N
SMILES: NC1=NNC(=N1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)