Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1,2,4-triazole-5-carboxylic acid hydrate, 98%, Thermo Scientific™

Catalog No. AAA1469030
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1469030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1469030 Supplier Thermo Scientific Chemicals Supplier No. A1469030

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Amino-1,2,4-triazole-5-carboxylic acid hydrate (AmTAZAc) has been used in the synthesis of new three-dimensional metal-organic framework, [ZnF(AmTAZ)] solvents.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304655-78-5
Molecular Formula C3H4N4O2
Molecular Weight (g/mol) 128.09
MDL Number MFCD00010620 MFCD00179325 MFCD00225624
InChI Key MVRGLMCHDCMPKD-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazole-5-carboxylic acid hydrate, 1h-1,2,4-triazole-3-carboxylicacid, 5-amino-, hydrate 1:1, 3-amino-5-carboxy-1h-1,2,4-triazole hydrate, 5-amino-2h-1,2,4-triazole-3-carboxylic acid hydrate, 3-amino-1,2,4-triazole-5-carboxylic acid hemihydrate, 3-amino-1h-1,2,4-triazole-5-carboxylic acid hydrate, 3-amino-1h-1,2,4-triazole-5-carboxylic acid, oxamethane, 5-amino-1 h-1,2,4-triazole-3-carboxylic acid, 5-amino-4 h-1,2,4 triazole-3-carboxylic acid
PubChem CID 16211818
IUPAC Name 3-amino-1H-1,2,4-triazole-5-carboxylic acid;hydrate
SMILES NC1=NNC(=N1)C(O)=O

Specifications

Melting Point ∼182°C (decomposition)
Quantity 250 g
Beilstein 124734
Solubility Information Slightly soluble in water.
Formula Weight 128.09 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 3-Amino-1,2,4-triazole-5-carboxylic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-868-1
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.