CAS RN 304671-92-9
IUPAC Name:
2-amino-3-(4-aminophenyl)propanoic acid hydrate
Synonyms:
P-aminophenylalanine hydrate
Molecular Weight (g/mol):
198.22
Molecular Formula:
C9H14N2O3
InChi Key:
LPKPWQCBWORKIP-UHFFFAOYNA-N
SMILES:
O.NC(CC1=CC=C(N)C=C1)C(O)=O
1
–
1
of
1
results
1
4-Amino-L-phenylalanine Monohydrate 98.0+%, TCI America™
CAS: 304671-92-9 Molecular Formula: C9H14N2O3 Molecular Weight (g/mol): 198.222 MDL Number: MFCD00150889 InChI Key: LPKPWQCBWORKIP-QRPNPIFTSA-N Synonym: H-Phe(4-NH2)-OH PubChem CID: 21117064 IUPAC Name: (2S)-2-amino-3-(4-aminophenyl)propanoic acid;hydrate SMILES: C1=CC(=CC=C1CC(C(=O)O)N)N.O