Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-L-phenylalanine Monohydrate 98.0+%, TCI America™
SDP

Catalog No. A26511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26511G Supplier TCI America Supplier No. A26511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 304671-92-9
Molecular Formula C9H14N2O3
Molecular Weight (g/mol) 198.222
MDL Number MFCD00150889
InChI Key LPKPWQCBWORKIP-QRPNPIFTSA-N
Synonym H-Phe(4-NH2)-OH
PubChem CID 21117064
IUPAC Name (2S)-2-amino-3-(4-aminophenyl)propanoic acid;hydrate
SMILES C1=CC(=CC=C1CC(C(=O)O)N)N.O

Specifications

Melting Point 240°C
Color White
Quantity 1 g
Formula Weight 180.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-L-phenylalanine Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.