accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSONa

CAS RN 304675-78-3

IUPAC Name: sodium (1-oxidopyridin-1-ium-2-yl)sulfanide
Synonyms: pyrithione sodium
Molecular Weight (g/mol): 149.14
Molecular Formula: C5H4NNaOS
InChi Key: WNGMMIYXPIAYOB-UHFFFAOYSA-M
SMILES: [Na+].[O-][N+]1=CC=CC=C1[S-]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results