accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30478-88-7

IUPAC Name: 3-bromonaphthalen-2-ol
Synonyms: 3-bromonaphthalen-2-ol
Molecular Weight (g/mol): 223.07
Molecular Formula: C10H7BrO
InChi Key: PQVIOPAWVAOHOA-UHFFFAOYSA-N
SMILES: OC1=CC2=CC=CC=C2C=C1Br
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)