Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-naphthol 96.0+%, TCI America™
SDP

Catalog No. B31275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31275G 5 g
B31271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31275G Supplier TCI America Supplier No. B31275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30478-88-7
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.07
MDL Number MFCD10000951
InChI Key PQVIOPAWVAOHOA-UHFFFAOYSA-N
PubChem CID 45943
IUPAC Name 3-bromonaphthalen-2-ol
SMILES OC1=CC2=CC=CC=C2C=C1Br

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 5 g
Formula Weight 223.07
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-naphthol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.