accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3048-64-4

IUPAC Name: 5-ethenylbicyclo[2.2.1]hept-2-ene
Synonyms: vinylnorbornene
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: INYHZQLKOKTDAI-UHFFFAOYNA-N
SMILES: C=CC1CC2CC1C=C2
Boiling Point 
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (5)