Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Vinyl-2-norbornene, mixture of endo and exo

Catalog No. 148679500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-867-9500ML 500 mL
14-867-925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-867-9500ML Supplier Sigma Aldrich Supplier No. 148679500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5-Vinyl-2-norbornene, mixture of endo and exo

Specifications

CAS 3048-64-4
Melting Point -80°C (lit.)
Density 0.841 g/mL (at 25°C (literature))
Boiling Point 141°C (lit.)
Molecular Formula C9H12
Linear Formula C9H12
Refractive Index n20/D 1.481 (literature)
MDL Number MFCD00080691
Quantity 500 mL
Molecular Weight (g/mol) 120.19
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.