accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 305372-39-8

IUPAC Name: N-[2-(2-{2-[3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido]ethoxy}ethoxy)ethyl]-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
Synonyms: N-[2-(2-{2-[3-(2,5-dioxopyrrol-1-yl)propanamido]ethoxy}ethoxy)ethyl]-5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamide
Molecular Weight (g/mol): 525.62
Molecular Formula: C23H35N5O7S
InChi Key: UDPCXVIKHFVPSZ-UHFFFAOYNA-N
SMILES: O=C(CCCCC1SCC2NC(=O)NC12)NCCOCCOCCNC(=O)CCN1C(=O)C=CC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results