Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Biotinyl-N'-(3-maleimidopropionyl)-3,6-dioxaoctane-1,8-diamine 90.0+%, TCI America™
SDP

Catalog No. B317450MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B317450MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B317450MG Supplier TCI America Supplier No. B317450MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 305372-39-8
Molecular Formula C23H35N5O7S
Molecular Weight (g/mol) 525.621
InChI Key UDPCXVIKHFVPSZ-HOIFWPIMSA-N
Synonym N-[2-[2-[2-[(3-Maleimidopropionyl)amino]ethoxy]ethoxy]ethyl]biotinamide, Maleimide-PEG2-Biotin
PubChem CID 11168457
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-[3-(2,5-dioxopyrrol-1-yl)propanoylamino]ethoxy]ethoxy]ethyl]pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCOCCOCCNC(=O)CCN3C(=O)C=CC3=O)NC(=O)N2

Specifications

Color White-Yellow
Quantity 50 mg
Formula Weight 525.62
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Biotinyl-N′-(3-maleimidopropionyl)-3,6-dioxaoctane-1,8-diamine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.