CAS RN 305372-78-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Wnt Pathway Inhibitor XI, CCT036477
The Wnt Pathway Inhibitor XI, CCT036477, also referenced under CAS 305372-78-5, controls the biological activity of Wnt Pathway. This small molecule/inhibitor is primarily used for Cancer applications.