Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Wnt Pathway Inhibitor XI, CCT036477

Catalog No. 6816745MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-167-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-167-45MG Supplier MilliporeSigma™ Supplier No. 6816745MG
Add to Cart
Add to Cart

The Wnt Pathway Inhibitor XI, CCT036477, also referenced under CAS 305372-78-5, controls the biological activity of Wnt Pathway. This small molecule/inhibitor is primarily used for Cancer applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by 1H NMR
CAS 305372-78-5
Molecular Formula C21H18ClN3
Formula Weight 347.84g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.