accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(S)OOOO

CAS RN 3054-07-7

IUPAC Name: (2S)-2-azaniumyloctanedioate
Synonyms: (2S)-2-ammoniooctanedioate
Molecular Weight (g/mol): 188.2
Molecular Formula: C8H14NO4
InChi Key: YOFPFYYTUIARDI-LURJTMIESA-M
SMILES: [NH3+][C@@H](CCCCCC([O-])=O)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results