Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Aminosuberic Acid 95.0+%, TCI America™
SDP

Catalog No. A12795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12795G Supplier TCI America Supplier No. A12795G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3054-07-7
Molecular Formula C8H14NO4
Molecular Weight (g/mol) 188.20
MDL Number MFCD00063122
InChI Key YOFPFYYTUIARDI-LURJTMIESA-M
Synonym DL-2-Aminooctanedioic Acid
PubChem CID 77937
IUPAC Name 2-aminooctanedioic acid
SMILES C(CCC(C(=O)O)N)CCC(=O)O

Specifications

Melting Point 240°C
Color White
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-2-Aminosuberic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.