accessibility menu, dialog, popup

UserName

Viewing profile as user:
NO

CAS RN 3055-86-5

IUPAC Name: 3-phenoxypropanenitrile
Synonyms: β-phenoxypropionitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: IXAUFLAHUXISCH-UHFFFAOYSA-N
SMILES: N#CCCOC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results