Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenoxypropionitrile 98.0+%, TCI America™
SDP

Catalog No. P14265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14265G Supplier TCI America Supplier No. P14265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3055-86-5
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00013822
InChI Key IXAUFLAHUXISCH-UHFFFAOYSA-N
PubChem CID 76456
IUPAC Name 3-phenoxypropanenitrile
SMILES N#CCCOC1=CC=CC=C1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenoxypropionitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.