CAS RN 3055-95-6
IUPAC Name:
3,6,9,12,15-pentaoxaheptacosan-1-ol
Synonyms:
laureth-5
Molecular Weight (g/mol):
406.6
Molecular Formula:
C22H46O6
InChi Key:
LAPRIVJANDLWOK-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCOCCOCCOCCOCCOCCO
1
–
1
of
1
results
1
Pentaethylene Glycol Monododecyl Ether 98.0+%, TCI America™
CAS: 3055-95-6 Molecular Formula: C22H46O6 Molecular Weight (g/mol): 406.60 MDL Number: MFCD00043064 InChI Key: LAPRIVJANDLWOK-UHFFFAOYSA-N PubChem CID: 18281 IUPAC Name: 3,6,9,12,15-pentaoxaheptacosan-1-ol SMILES: CCCCCCCCCCCCOCCOCCOCCOCCOCCO