Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaethylene Glycol Monododecyl Ether 98.0+%, TCI America™
SDP

Catalog No. P03725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P03725G Supplier TCI America Supplier No. P03725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3055-95-6
Molecular Formula C22H46O6
Molecular Weight (g/mol) 406.60
MDL Number MFCD00043064
InChI Key LAPRIVJANDLWOK-UHFFFAOYSA-N
PubChem CID 18281
IUPAC Name 3,6,9,12,15-pentaoxaheptacosan-1-ol
SMILES CCCCCCCCCCCCOCCOCCOCCOCCOCCO

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 406.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Pentaethylene Glycol Monododecyl Ether
EINECSNumber (7)-0097
RTECSNumber SB0333332
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.