accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOOH

CAS RN 3055-96-7

IUPAC Name: 3,6,9,12,15,18-hexaoxatriacontan-1-ol
Synonyms: 3,6,9,12,15,18-hexaoxatriacontan-1-ol
Molecular Weight (g/mol): 450.66
Molecular Formula: C24H50O7
InChi Key: OJCFEGKCRWEVSN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results