Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexaethylene Glycol Monododecyl Ether 97.0+%, TCI America™
SDP

Catalog No. H04441ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04441ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04441ML Supplier TCI America Supplier No. H04441ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3055-96-7
Molecular Formula C24H50O7
Molecular Weight (g/mol) 450.657
MDL Number MFCD00042660
InChI Key OJCFEGKCRWEVSN-UHFFFAOYSA-N
PubChem CID 18282
IUPAC Name 2-[2-[2-[2-[2-(2-dodecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCO

Specifications

Color White
Quantity 1 mL
Formula Weight 450.66
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Hexaethylene Glycol Monododecyl Ether
EINECSNumber (7)-0097
RTECSNumber MP4780500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.