accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3055-97-8

H3COOOOOOOOH

CAS RN 3055-97-8

IUPAC Name: 3,6,9,12,15,18,21-heptaoxatritriacontan-1-ol
Synonyms: 3,6,9,12,15,18,21-heptaoxatritriacontan-1-ol
Molecular Weight (g/mol): 494.71
Molecular Formula: C26H54O8
InChi Key: DWHIUNMOTRUVPG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results