Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptaethylene Glycol Monododecyl Ether, TCI America™
SDP

Catalog No. H04451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04451G Supplier TCI America Supplier No. H04451G

Chemical Identifiers

CAS 3055-97-8
Molecular Formula C26H54O8
Molecular Weight (g/mol) 494.71
MDL Number MFCD00043065
InChI Key DWHIUNMOTRUVPG-UHFFFAOYSA-N
PubChem CID 76459
IUPAC Name 2-[2-[2-[2-[2-[2-(2-dodecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 494.71
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Heptaethylene Glycol Monododecyl Ether
EINECSNumber (7)-0097
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.