accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNNONNH3COHN

CAS RN 3056-33-5

IUPAC Name: N-(9-acetyl-6-oxo-6,9-dihydro-3H-purin-2-yl)acetamide
Synonyms: N2,9-diacetylguanine
Molecular Weight (g/mol): 235.2
Molecular Formula: C9H9N5O3
InChi Key: GILZZWCROUGLIS-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(C)=O
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)