Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N2,9-Diacetylguanine 95.0+%, TCI America™
SDP

Catalog No. D360425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D360425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D360425G Supplier TCI America Supplier No. D360425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3056-33-5
Molecular Formula C9H9N5O3
Molecular Weight (g/mol) 235.20
MDL Number MFCD00142116
InChI Key GILZZWCROUGLIS-UHFFFAOYSA-N
Synonym n2,9-diacetylguanine, n,9-diacetylguanine, n 2 ,9-diacetylguanine, 2,9-diacetylguanine, 2,9-diacetateguanine, n-9-acetyl-6-oxo-6,9-dihydro-1h-purin-2-yl acetamide, unii-vuw2b9yr95, 2,9-diacetyl guanine, vuw2b9yr95, n-9-acetyl-6-oxo-3h-purin-2-yl acetamide
PubChem CID 76461
IUPAC Name N-(9-acetyl-6-oxo-6,9-dihydro-3H-purin-2-yl)acetamide
SMILES CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(C)=O

Specifications

Melting Point 285°C
Quantity 25 g
Formula Weight 235.20
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N2,9-Diacetylguanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.