accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3H3CCH3

CAS RN 3056-64-2

IUPAC Name: 4-tert-butylphenyl acetate
Synonyms: P-tert-butylphenyl acetate
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: FSALNWWFUMHOAU-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results