Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphenyl Acetate 97.0+%, TCI America™
SDP

Catalog No. B09625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09625ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B09625ML Supplier TCI America Supplier No. B09625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3056-64-2
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00026308
InChI Key FSALNWWFUMHOAU-UHFFFAOYSA-N
Synonym Acetic Acid 4-tert-Butylphenyl Ester
PubChem CID 76463
IUPAC Name (4-tert-butylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C)(C)C

Specifications

Color Yellow
Boiling Point 245°C
Quantity 5 mL
Formula Weight 192.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylphenyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.